Please login to your online account to display your discounted pricing

Thermo Scientific™ 384-well Microarray Plate


384-well microarray plate designed for low volume sample storage.

Manufacturer: thermo scientific™  AB1055

 View more versions of this product

Catalog No. AB-1055

  • / Case of 50 Each
 

Description & Specifications

Specifications

Color Natural
For Use With (Application) For use with the widest range of sealing options
Material Polypropylene
Max. RCF 5000xG
No. of Wells 384
Type 384-Well Microarray Plate
Volume (Metric) Well 70μL
Well Design Round