Please login to your online account to display your discounted pricing

Thermo Scientific™ 384-well Microarray Plate

384-well microarray plate designed for low volume sample storage.

$787.03 - $789.27

Specifications

Color Natural
Type 384-Well Microarray Plate
For Use With (Application) For use with the widest range of sealing options
Material Polypropylene
Max. RCF 5000xG
View More Specs
Products
Catalog Number Mfr. No. Volume (Metric) Well Price Quantity    

AB-1055

 
thermo scientific™
AB1055
70μL Case of 50 Each for $787.03

AB-1056

 
thermo scientific™
AB1056
55μL Case of 50 Each for $789.27
Description & Specifications

Specifications

Color Natural
Type 384-Well Microarray Plate
For Use With (Application) For use with the widest range of sealing options
Material Polypropylene
Max. RCF 5000xG
No. of Wells 384

Storage and Assay Plates are manufactured and in a class 100,000 clean room environment to ensure that each plate is contamination free. Plates are made using virgin medical grade polypropylene and have highly polished surfaces making them low binding and chemically resistant. Rigid side walls and industry standard footprint make plates suitable for automation and robotic handling, as well as for low throughput applications.

Thermo Scientific Storage and Assay Plates are manufactured and in a class 100,000 clean room environment to ensure that each plate is contamination free. Plates are made using virgin medical grade polypropylene and have highly polished surfaces making them low binding and chemically resistant. Rigid side walls and industry standard footprint make plates suitable for automation and robotic handling, as well as for low throughput applications.
Highlights
• Designed for optimal sample recovery
• Each well specifically designed with an independent sealing rim to prevent cross-contamination
• Ideally suited for use with the widest range of sealing options
• Improved sample recovery and decreased dead volume
• Available with conical (70 μL maximum volume) or v-bottom (55 μL maximum volume) wells

Compatible Sealing Film:Adhesive, Heat
> Hazardous:No
>Max Centrifugal Force:5000 g
> Max Vol (Cap Seal):n/a
> Max Vol (Film Seal):70 μL (AB-1055) or 55 μL (AB-1056)
> Max Vol (Mat Seal):30 μL (AB-1055) or 25 μL (AB-1056)
> Storage Condition:Ambient

Certified free from human DNA, RNases, and DNases.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.