Thermo Scientific™ 2.2 mL Storage Plate
2.2 mL deepwell storage plates for the culture or storage of large samples.
$728.88 - $1,118.06
Products ${productFamilyLength}
Catalog Number Mfr. No. Well Design Price Quantity  
Catalog Number Mfr. No. Well Design Price Quantity  
AB-0932B Thermo Scientific™
AB0932B
Square Case of 50 Each for $728.88
AB0788 Thermo Scientific™
AB0788
Round (U) Case for $1,118.06
Specifications
Description
Streamline your molecular biology applications with Thermo Scientific storage and assay plates. All plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
Streamline your molecular biology applications with Thermo Scientific storage and assay plates. All plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
Highlights
• Designed for optimal sample recovery
• Each square well is specifically designed with an independent sealing rim to prevent cross-contamination
• U-bottom (AB-0661) to aid resuspension or conical bottom (AB-0932) for ease of sample retrieval
Compatible Sealing Film:Adhesive Heat
> Hazardous:No
>Max Centrifugal Force:5000 g
> Max Vol (Film Seal):2.2 mL
> Max Vol (Mat Seal):1.8 mL
> Storage Condition:Ambient
Certifications
Certified free from human DNA, RNase, and DNases.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.