Please login to your online account to display your discounted pricing
Thermo Scientific™ 0.2 mL Strip Tubes
Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.
Manufacturer: thermo scientific™  AB-0382
 View more versions of this product
Catalog No. AB-0382
  • / Case of 8 Each
Description & Specifications
Specifications
Type 8-Cap strip
Cap Type Domed
Certifications/Compliance Certified free from human DNA, RNases, and DNases
Closure Color Natural
Color --
Material Polypropylene
Quantity 10 packs of 12 strips
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.
Highlights
  • Compatible with 0.2 mL thermal cycler blocks
  • 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays