Please login to your online account to display your discounted pricing

Thermo Scientific™ 0.2 mL Strip Tubes


Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.

Manufacturer: thermo scientific™  AB-0266/B

 View more versions of this product

Catalog No. AB-0266B

  • / Case of 250 Each
 

Description & Specifications

Specifications

Type 8-Tube strip
Cap Type Domed
Certifications/Compliance Certified free from human DNA, RNases, and DNases
Color Blue
Material Polypropylene
Quantity 250 strips
Shape V-Bottom
Capacity (Metric) 0.2mL

0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.

Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.

Highlights

  • Compatible with 0.2 mL thermal cycler blocks
  • 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays