Please login to your online account to display your discounted pricing
Thermo Scientific™ 0.2 mL Strip Tubes
Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.
Manufacturer: thermo scientific™  AB0452G
 View more versions of this product
Catalog No. AB-0452G
  • / Case of 120 Each
Description & Specifications
Specifications
Type 8-Tube strip
Quantity 10 packs of 12 strips
Cap Type None
Shape V-Bottom
Capacity (Metric) 0.2mL
Color Green
Material Polypropylene
Certifications/Compliance Certified free from human DNA, RNases, and DNases
0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.
Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.