Please login to your online account to display your discounted pricing

Thermo Scientific™ 0.2 mL Individual Tubes


0.2 mL individual tubes with attached flat or dome snap shut caps, available in a variety of colors

Manufacturer: thermo scientific™  AB-0337/B

 View more versions of this product

Catalog No. AB-0337B

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Type Thin Wall PCR Tube
Cap Type Domed
Certifications/Compliance RNase, Dnase and human DNA free
Color Blue
Material Polypropylene
Volume (Metric) 0.2mL
Quantity 1000 Case
Format Tube
For Use With (Equipment) 0.2mL thermal cycler blocks for PCR and sample storage applications
Capacity (Metric) 0.2mL

0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.

The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when closed

Integral 'snap shut' cap
• Available with flat or domed caps
Applications
• PCR
• Sample storage

Hazardous:No
>Storage Condition:Ambient

PCR; Sample storage