Fisherbrand™ Thermal Mailer with Tape and Zipper Closure
Fisherbrand Thermal Mailer with Tape and Zipper Closure is ideal for short trips and whenever maintaining the cold chain of pharmaceuticals or laboratory products as required.
$436.67 - $516.73
Specifications
Closure Type White Tape, Dual closure with leakproof zipper inside and exterior self-seal strip
Color Metalized PET Silver Exterior
Material Outer Layer: Metalized PET laminated to PE, Middle Layer: Open Cell PU Foam, Inner Layer: White LDPE (FDA approved for incidental food contact).
Quantity 25
Type Thermal Mailer
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
07-100-105 View Documents Fisherbrand™
KG1216PL05UXUM25
Outside: 12 x 16 in.; Inside: 11 x 14 in. Case of 25 Each for $516.73
07-100-104 View Documents Fisherbrand™
KG0912PL05UXUS25
Outside: 9 x 12 in.; Inside: 8 x 10 in. Case of 25 Each for $436.67
07-100-106 View Documents Fisherbrand™
KG1520PL05UXUL25
Outside: 15 x 20 in.; Inside: 14 x 18 in. Case of 20 Each for $459.33
Description
Description
Features:
  • Thick padded mailers cushion and insulate up to four hours
  • Dual closure with leakproof zipper inside and exterior self-seal strip
  • Very thick foam (0.5in.) with metallized poly exterior
  • Made in USA
Order Info
Case quantity only 25 per case
Warranty
Warranty against defects in material or workmanship.
Specifications
Specifications
White Tape, Dual closure with leakproof zipper inside and exterior self-seal strip
Outer Layer: Metalized PET laminated to PE, Middle Layer: Open Cell PU Foam, Inner Layer: White LDPE (FDA approved for incidental food contact).
Thermal Mailer
Metalized PET Silver Exterior
25