missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Therapak™ Coleman™ Excursion™ Handle Coolers
For safe transport of contents
Manufacturer: Therapak™ TR56229G
Description
- Generous foam insulation, along with hinged lids and lid-locking handles
- Available in red and blue color
- Capacity: 9 to 16 qt. (8.51 to 15.14L)
Specifications
15.14 L | |
Blue | |
Excursion Cooler | |
Hinged |