missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Therapak™ Coleman™ Molded Handle Coolers
With 0% ozone depleting materials
Manufacturer: Therapak™ TR56240G
Description
- Made of environmentally friendly insulation
- Molded handles for easy carrying
- Removable lid
Specifications
34 L | |
Coleman Cooler | |
36 qt. |
Blue | |
Removable | |
0% Ozone depleting materials, molded handle |