Learn More
Theophylline-7-acetic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1147314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Theophylline-7-acetic acid | |
268°C to 272°C | |
MFCD00022832 | |
279221 | |
acefylline, theophylline-7-acetic acid, acephylline, theophyllineacetic acid, carboxymethyltheophylline, 7-carboxymethyl theophylline, 7-theophyllinylacetic acid, theophyllin-7-ylacetic acid, 7-theophyllinessigsaeure, 7-theophyllineacetic acid | |
[Na+].CN1C2=C(N(CC([O-])=O)C=N2)C(=O)N(C)C1=O | |
260.19 | |
238.2 |
652-37-9 | |
C9H9N4NaO4 | |
25 g | |
14,24 | |
MSFVZSOKOXZSME-UHFFFAOYSA-M | |
2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetic acid | |
69550 | |
98% |
Chemical Identifiers
652-37-9 | |
260.19 | |
MSFVZSOKOXZSME-UHFFFAOYSA-M | |
69550 | |
[Na+].CN1C2=C(N(CC([O-])=O)C=N2)C(=O)N(C)C1=O |
C9H9N4NaO4 | |
MFCD00022832 | |
acefylline, theophylline-7-acetic acid, acephylline, theophyllineacetic acid, carboxymethyltheophylline, 7-carboxymethyl theophylline, 7-theophyllinylacetic acid, theophyllin-7-ylacetic acid, 7-theophyllinessigsaeure, 7-theophyllineacetic acid | |
2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 211-490-2
RTECSNumber : UO7451300
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only