Please login to your online account to display your discounted pricing

Themis Rat anti-Mouse, PE, Clone: 1TMYS, eBioscience™

Rat Monoclonal Antibody

$187.05 - $187.05

Specifications

Antigen Themis
Clone 1TMYS
Host Species Rat
Gene Alias Gasp
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122346

 
life technologies
LS12591880
25μg Each for $187.05
Description & Specifications

Specifications

Antigen Themis
Clone 1TMYS
Host Species Rat
Gene Alias Gasp
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This 1TMYS monoclonal antibody reacts with mouse Thymocyte-expressed molecule involved in selection (Themis), also known as Grb2-associating protein (Gasp). Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling.

There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing. The 1TMYS antibody recognizes isoforms 1, 2, 3, and 4, but does not recognize isoform 5.