Please login to your online account to display your discounted pricing

Thapsigargin, 97%, ACROS Organics™


Quantity 1mg

Manufacturer: thermo fisher scientific  328570010

 View more versions of this product

Catalog No. AC328570010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. Wear respiratory protection. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Assay ≥96.0%
Physical Form Film
Formula Weight 650.76
Quantity 1mg
Color Colorless
Merck Index 15, 9421
UN Number 2811
MDL Number MFCD00083511
CAS 67526-95-8
Chemical Name or Material Thapsigargin
Assay Percent Range 97%
Molecular Formula C34H50O12
Health Hazard 1 Danger

Specifications

Chemical Name or Material Thapsigargin
CAS 67526-95-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C34H50O12
Formula Weight 650.76
MDL Number MFCD00083511
Accessories

Danger

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. Wear respiratory protection. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled.

SDS & Safety Documents