Please login to your online account to display your discounted pricing

Thallium Rod, 12.7mm (0.5in) dia., 99.9% (Metals basis), Alfa Aesar™

Form: Approx. 225g/150mm

Chemical Identifiers

Chemical Name or Material Thallium
CAS 7440-28-0
Physical Form Rod, 12.7mm (0.5 in.) dia.
Assay Percent Range 99.9% (Metals basis)
MDL Number MFCD00134063
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA123520Y

  SDS
alfa aesar
123520Y
75mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12352DO

  SDS
alfa aesar
12352DO
150mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 75mm
Physical Form Rod, 12.7mm (0.5 in.) dia.
EINECS Number 231-138-1
Assay Percent Range 99.9% (Metals basis)
Health Hazard 1 H300-H330-H373-H413: Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.
Appearance Rod
MDL Number MFCD00134063
CAS 7440-28-0
Chemical Name or Material Thallium
Odor Odorless
Safety and Handling

H300-H330-H373-H413: Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.