missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thallium Rod, 12.7mm (0.5in) dia., 99.9% (Metals basis), Alfa Aesar™

Form: Approx. 225g/150mm

Manufacturer:  Alfa Aesar 12352DO

 View more versions of this product

Catalog No. AA12352DO



Specifications

Specifications

Thallium
7440-28-0
99.9% (Metals basis)
MFCD00134063
BKVIYDNLLOSFOA-UHFFFAOYSA-N
thallium
204.38
CHEBI:30440
150mm
Rod
Tl
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
[Tl]
Odorless
5359464
Rod, 12.7mm (0.5 in.) dia.

Chemical Identifiers

7440-28-0
204.38
BKVIYDNLLOSFOA-UHFFFAOYSA-N
5359464
thallium
Tl
MFCD00134063
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
CHEBI:30440
[Tl]
Safety and Handling

Safety and Handling

H300-H330-H373-H413: Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.

EINECSNumber : 231-138-1

Documents