missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thallium rod, 12.7mm (0.5 in.) dia., 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 12714DO

 View more versions of this product

Catalog No. AA12714DO



Specifications

Specifications

Thallium rod
150mm
7440-28-0
Tl
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
[Tl]
Odorless
5359464
Rod
≈225g/150mm
12.7mm (0.5 in.) dia.
(metals basis)
MFCD00134063
BKVIYDNLLOSFOA-UHFFFAOYSA-N
thallium
204.38
CHEBI:30440
99.999%

Chemical Identifiers

7440-28-0
204.38
BKVIYDNLLOSFOA-UHFFFAOYSA-N
5359464
thallium
Tl
MFCD00134063
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
CHEBI:30440
[Tl]
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents