Learn More
Alfa Aesar™ Thallium, plasma standard solution, Specpure™, Tl 1000μg/mL
Manufacturer: Alfa Aesar™ 13851AP
Specifications
Thallium plasma standard solution | |
MFCD00134063 | |
Tl in 5% HNO3 | |
Specpure™ |
Tl 1000μg/mL | |
UN3264 | |
Liquid | |
500mL |
Chemical Identifiers
MFCD00134063 |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
TSCA : TSCA