Please login to your online account to display your discounted pricing

Alfa Aesar™ Thallium(III) trifluoroacetate, tech.

Chemical Identifiers

Chemical Name or Material Thallium (III) trifluoroacetate
CAS 23586-53-0
Purity Grade Technical Grade
Molecular Formula Tl(OOCCF3)3
Formula Weight 543.42
MDL Number MFCD00000414
UN Number UN1707
Synonym Thallic trifluoroacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4364309

  SDS
alfa aesar™
4364309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4364318

  SDS
alfa aesar™
4364318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4364304

  SDS
alfa aesar™
4364304
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Thallium (III) trifluoroacetate
CAS 23586-53-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
TSCA TSCA
Purity Grade Technical Grade
Physical Form Powder
Molecular Formula Tl(OOCCF3)3
Formula Weight 543.42
MDL Number MFCD00000414
UN Number UN1707
Synonym Thallic trifluoroacetate
Sensitivity Hygroscopic
Melting Point 213°C (dec.)
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.