missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thallium ICP Standard, 1000 ppm Tl in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PTL1KN100
Specifications
Thallium ICP Standard | |
Natural LDPE Bottle | |
93.0,4.76,0.09 | |
Colorless | |
Tl | |
[Tl] | |
204.38 | |
CHEBI:30440 |
7440-28-0,7697-37-2,7732-18-5 | |
100 mL | |
97.0,4.95,0.1 | |
1,000ppm Tl | |
BKVIYDNLLOSFOA-UHFFFAOYSA-N | |
thallium | |
5359464 | |
1 mL = 1 mg Tl |
Chemical Identifiers
7440-28-0 | |
204.38 | |
5359464 | |
thallium |
Tl | |
BKVIYDNLLOSFOA-UHFFFAOYSA-N | |
CHEBI:30440 | |
[Tl] |