Please login to your online account to display your discounted pricing

Alfa Aesar™ Thallium(I) sulfate, Puratronic™, 99.9995% (metals basis)

Chemical Identifiers

Chemical Name or Material Thallium(I) sulfate
CAS 7446-18-6
Purity Grade Puratronic™
Formula Weight 504.80g/mol
MDL Number MFCD00011278
UN Number UN1707
Merck Index 14,9269
Synonym Thallous sulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4021506

 
alfa aesar™
4021506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4021514

 
alfa aesar™
4021514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 6.77g/mL
Odor Odorless
Quantity 5g
Boiling Point (dec.)
Melting Point 632°C
Quantity 5g
Chemical Name or Material Thallium(I) sulfate
CAS 7446-18-6
Purity Grade Puratronic™
Physical Form Crystalline
Assay 99.9995%
Assay Percent Notes (metals basis)
Formula Weight 504.80g/mol
MDL Number MFCD00011278
UN Number UN1707
Merck Index 14,9269
Synonym Thallous sulfate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H372-H315
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P260-P280-P301+P310-P362-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P280-P301+P310-P362-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H372-H315
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.