Please login to your online account to display your discounted pricing

Alfa Aesar™ Thallium(I) neodecanoate, (Tl 52.5-55.5%)

Chemical Identifiers

Chemical Name or Material Thallium(I) neodecanoate
Formula Weight 375.61
MDL Number MFCD00070467
UN Number UN1707
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3962609

  SDS
alfa aesar™
3962609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3962618

  SDS
alfa aesar™
3962618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 375.61
Physical Form Solid
Concentration Tl 52.5 to 55.5%
Health Hazard 1 Danger
Quantity 10g
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
Health Hazard 2 GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00070467
UN Number UN1707
Chemical Name or Material Thallium(I) neodecanoate
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.