Learn More
Thallium(I) hexafluorophosphate(V), 97% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 3962706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thallium(I) hexafluorophosphate(V) | |
Crystalline | |
F6PTl | |
UN1707 | |
Soluble in water. | |
F[P-](F)(F)(F)(F)F.[Tl+] | |
349.344 | |
349.34 |
60969-19-9 | |
5 g | |
MFCD00049807 | |
thallium hexafluorophosphate, tlpf6, thallium i hexafluorophosphate, thallium 1+ hexafluorophosphate, thallium 1+ hexafluoro-$l^ 5-phosphanuide, $l^ 1-thallanylium hexafluoro-$l^ 5-phosphanuide | |
FRZBCOUMLHRKRT-UHFFFAOYSA-N | |
thallium(1+);hexafluorophosphate | |
10904204 | |
≤97% |
Chemical Identifiers
60969-19-9 | |
349.344 | |
FRZBCOUMLHRKRT-UHFFFAOYSA-N | |
10904204 | |
F[P-](F)(F)(F)(F)F.[Tl+] |
F6PTl | |
MFCD00049807 | |
thallium hexafluorophosphate, tlpf6, thallium i hexafluorophosphate, thallium 1+ hexafluorophosphate, thallium 1+ hexafluoro-$l^ 5-phosphanuide, $l^ 1-thallanylium hexafluoro-$l^ 5-phosphanuide | |
thallium(1+);hexafluorophosphate |
Safety and Handling
GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
H300+H330-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only