Please login to your online account to display your discounted pricing

Thallium(I) chloride, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  208881000

 View more versions of this product

Catalog No. AC208881000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥98.5%
Formula Weight 239.82
Quantity 100g
Color White to off-white
EINECS Number 232-241-4
TSCA TSCA
Density 7g/mL
RTECS Number XG4200000
Merck Index 15, 9409
Packaging Glass bottle
Linear Formula TICl
MDL Number MFCD00011274
CAS 7791-12-0
Synonym Thallous chloride
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Thallium(I) chloride
Name Note 99%
Assay Percent Range 99%
Molecular Formula ClTl
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.

Specifications

Chemical Name or Material Thallium(I) chloride
Name Note 99%
CAS 7791-12-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula ClTl
Linear Formula TICl
Formula Weight 239.82
MDL Number MFCD00011274
Synonym Thallous chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.

SDS & Safety Documents