Please login to your online account to display your discounted pricing

Thallium(I) chloride, 99%, ACROS Organics™

$404.60 - $404.60

Chemical Identifiers

Chemical Name or Material Thallium(I) chloride
Name Note 99%
CAS 7791-12-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula ClTl
Linear Formula TICl
Formula Weight 239.82
MDL Number MFCD00011274
Synonym Thallous chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC208880250

  SDS
thermo fisher scientific
208880250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC208881000

  SDS
thermo fisher scientific
208881000
Glass bottle 100g Each for $404.60
Description & Specifications

Specifications

Physical Form Powder
Assay ≥98.5%
Formula Weight 239.82
Quantity 25g
Color White to off-white
Merck Index 15, 9409
EINECS Number 232-241-4
TSCA TSCA
Density 7g/mL
RTECS Number XG4200000
Name Note 99%
Linear Formula TICl
MDL Number MFCD00011274
CAS 7791-12-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Thallium(I) chloride
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula ClTl
Synonym Thallous chloride
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.