Please login to your online account to display your discounted pricing

Alfa Aesar™ Thallium(I) acetate, Puratronic™, 99.9985% (metals basis)

Chemical Identifiers

Chemical Name or Material Thallium(I) acetate
CAS 563-68-8
Purity Grade Puratronic™
Formula Weight 263.42
MDL Number MFCD00013045
UN Number UN1707
Beilstein 3693914
Merck Index 14,9257
Synonym Thallous acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1110222

  SDS
alfa aesar™
1110222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1110214

  SDS
alfa aesar™
1110214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 263.42
Chemical Name or Material Thallium(I) acetate
TSCA TSCA
Odor Odorless
Purity Grade Puratronic™
Physical Form Powder
Assay 99.9985%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00013045
CAS 563-68-8
UN Number UN1707
Beilstein 3693914
Merck Index 14,9257
Synonym Thallous acetate
Melting Point 131°C
Quantity 100g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.