Please login to your online account to display your discounted pricing

Alfa Aesar™ Thallium(I) 2-ethylhexanoate, 99.8% (metals basis)

Chemical Identifiers

Chemical Name or Material Thallium(I) 2-ethylhexanoate
CAS 210578-56-6
Formula Weight 347.59
MDL Number MFCD00070466
UN Number UN1707
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3665904

 
alfa aesar™
3665904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3665909

 
alfa aesar™
3665909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3665918

 
alfa aesar™
3665918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 347.59
Chemical Name or Material Thallium(I) 2-ethylhexanoate
Physical Form Solid
Assay 99.8%
Assay Percent Notes (metals basis)
MDL Number MFCD00070466
CAS 210578-56-6
UN Number UN1707
Quantity 2g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.