missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thallium(I) 2,4-pentanedionate, 97+%

Manufacturer:  Alfa Aesar™ 7811303

 View more versions of this product

Catalog No. AA7811303



Specifications

Specifications

Thallium (I) 2,4-pentanedionate
C5H7O2Tl
UN1707
QGQYKYKTENVDSC-BJILWQEISA-M
[(E)-4-oxopent-2-en-2-yl]oxythallium
71311301
Crystalline
>134°C
25955-51-5
MFCD00013506
thallium i acetylacetonate, th acac, 2,4-pentanedione thallium i derivative, thallium 1+ 2e-4-oxopent-2-en-2-olate
CC(=CC(=O)C)O[Tl]
303.489
303.5
≥97%
1g

Chemical Identifiers

25955-51-5
303.489
QGQYKYKTENVDSC-BJILWQEISA-M
71311301
CC(=CC(=O)C)O[Tl]
C5H7O2Tl
MFCD00013506
thallium i acetylacetonate, th acac, 2,4-pentanedione thallium i derivative, thallium 1+ 2e-4-oxopent-2-en-2-olate
[(E)-4-oxopent-2-en-2-yl]oxythallium
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

SDS Documents