Please login to your online account to display your discounted pricing

Thallic Acetate 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  420751000

 View more versions of this product

Catalog No. AC420751000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 381.51
Quantity 100g
Color White to off-white
Beilstein 02, 96
Merck Index 15, 9406
EINECS Number 219-913-2
TSCA TSCA
Fieser 01,1150; 02,406; 03,286; 04,492; 05,655; 07,360
Chemical Name or Material Thallic acetate
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H9O6Tl
Linear Formula (CH3CO2)3Tl
MDL Number MFCD00008698
CAS 2570-63-0
Health Hazard 2 GHS H Statement:
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Acetic acid, thallium(III) salt
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Thallic acetate
Name Note 98%
CAS 2570-63-0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H9O6Tl
Linear Formula (CH3CO2)3Tl
Formula Weight 381.51
MDL Number MFCD00008698
Synonym Acetic acid, thallium(III) salt
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents