missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thallic acetate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC420750050

 View more versions of this product

Catalog No. AC420750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Thallic acetate
2570-63-0
C6H9O6Tl
MFCD00008698
01,1150; 02,406; 03,286; 04,492; 05,655; 07,360
Acetic acid, thallium(III) salt
381.51
Crystalline Powder
Authentic
White
5g
98%
98%
(CH3CO2)3Tl
02, 96
15, 9406
Solubility in water: decomposes
381.51
98%
Glass bottle
182°C

Chemical Identifiers

2570-63-0
381.51
Acetic acid, thallium(III) salt
C6H9O6Tl
MFCD00008698
Safety and Handling

Safety and Handling

GHS H Statement:
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-913-2

TSCA : TSCA

SDS Documents