missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ TGFBR2 Recombinant Human Protein DFS Item

Easily screen for kinase binding. Invitrogen™ TGFBR2 Recombinant Human Protein fusion protein consists of the catalytic domain of transforming growth factor beta receptor 2 (TGFBR2) and glutathione S-transferase (GST) tag. It is validated in the LanthaScreen™ Eu Kinase Binding Assay.

$606.00 - $8,500.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6122
View Documents
Invitrogen™
LSPV6122
10μg Each for $606.00
 
PR9154A
View Documents
Invitrogen™
LSPR9154A
100μg Each for $2,040.00
 
PV6123
View Documents
Invitrogen™
LSPV6123
1mg Each for $8,500.00
 
Description

Description

The LanthaScreen™ Eu Kinase Binding Assay is a TR-FRET assay that lets you measure inhibitor binding to non-activated kinases, perform simple off-rate measurements and makes simple the study of type III (allosteric) inhibitors.

The LanthatScreen™ Eu Kinase Binding Assay is easy to optimize and run. The assay uses a simple mix-and-read format with a single one-hour incubation and no development steps. Simultaneous binding of an Alexa Fluor™ conjugate or tracer” to a tagged kinase is detected by addition of a europium-labeled anti-tag antibody, in this case anti-GST. Simultaneous binding of the tracer and antibody to the kinase results in a high degree of fluorescence resonance energy transfer (FRET), while displacement of the tracer with a kinase inhibitor results in a loss of FRET.

  • Compatible and validated with the LanthaScreen™ Eu Kinase Binding Assay
  • Expressed in insect cells
  • GST tagged for use with Eu-labeled GST antibody
  • No special measures were taken to activate this kinase

TGFBR2 for the LanthaScreen Kinase Binding Assay
Transforming Growth Factor, beta receptor 2 (TGFBR2) is a member of the TGF-beta family of receptor Serine/Threonine kinases. It is involved in the regulation of cell proliferation. (TGFBR2 Aliases: AAT3, FAA3, MFS2, RIIC, LDS1B, LDS2B, TAAD2, TGFR-2, TGFbeta-RII).

This insect cell-expressed recombinant fusion protein of the TGFBR2 catalytic domain and GST tag has been validated in the LanthaScreen™ Eu Kinase Binding Assay with the Tracer 178 and Eu-labeled GST Antibody reagents.

Specifications

Specifications

Frozen
Human
GST Tag
Yes