missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human TGFBR1 (ALK5), GST Tag Recombinant Protein
Human TGFBR1 (ALK5), GST Tag Recombinant Protein
$786.00 - $2,645.00
Specifications
Accession Number | NP_004603.1 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 62.9 kDa |
Gene Alias | TGFBR1 (ALK5) |
KinaseFamily | STKR Kinase Family |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV5837
|
Invitrogen™
PV5837 |
10μg |
N/A
|
|
|||||
PR9075A
|
Invitrogen™
PR9075A |
100μg |
N/A
|
|
|||||
PV5838
|
Invitrogen™
PV5838 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Upon binding TGF-b, TGFBR2 dimerizes with and activates TGFBR1 via phosphorylation leading to downstream phosphorylation of SMADs by TGFBR1.
Specifications
NP_004603.1 | |
62.9 kDa | |
STKR Kinase Family | |
GST Tag | |
Ser⁄Thr Kinases: TKL Group | |
Baculovirus |
Kinase Assay | |
TGFBR1 (ALK5) | |
Human | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Dry Ice |