Please login to your online account to display your discounted pricing

TGF beta Rat anti-Human, Clone: eBioTB2F, eBioscience™

Rat Monoclonal Antibody

$129.05 - $390.05

Specifications

Antigen TGF beta
Clone eBioTB2F
Host Species Rat
Gene Alias TGFb, transforming growth factor
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122895

 
affymetrix
14994385
500μg Each for $390.05

5013903

 
affymetrix
14994381
50μg Each for $129.05
Description & Specifications

Specifications

Antigen TGF beta
Clone eBioTB2F
Host Species Rat
Gene Alias TGFb, transforming growth factor
Species Reactivity Human
Applications ELISA Capture
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The eBioTB2F antibody reacts with Transforming Growth Factor-beta (TGF-β), a pleiotropic cytokine, exists in five isoforms, known as TGF-β1-5. Homologies between isoforms range from 70-80% but no homology exists to TGF-α. TGF-β1 is ubiquitous and the most abundant form found in lymphoid organs, while other isoforms are expressed in a more restricted distribution. The biologically active state of all isoforms are disulfide-linked homodimers. The heat- and acid- stable monomeric subunits have a length of 112 amino acids. The isoforms of TGF-β arise by proteolytic cleavage of longer precursors; the isoforms are derived from the carboxyterminal ends of these precursors. Isoforms isolated from different species are evolutionarily closely conserved and have sequence identities on the order of 98%. Mature human, porcine, simian, chicken and bovine TGF-β1 are identical and differ from mouse TGF-β1 in a single amino acid. TGF-β1 is produced in very high levels by platelets. Other cellular sources of TGF-β1 include macrophages, lymphocytes, endothelial cells, chondrocytes, and leukemic cells. TGF-β1 secretion can be induced by steroids, retinoids, EGF, NGF, vitamin D3, and IL-1. Activities of TGF-β1 include inhibition of cell growth for inhibitor for normal and transformed epithelial cells, endothelial cells, fibroblasts, neurons, and lymphoid cells and other hematopoietic cell types. TGF-β1 inhibits the proliferation of T cells and NK cells and downregulates the activities of activated macrophages. TGF-β1 blocks the anti-tumor activity of IL-2 – bearing lymphokine-activated killer (LAK) cells.

Recently, TGF-β1 has been found to have a critical role in the development of regulatory T cells. Dendritic cells exposed to tumors have been reported to secrete TGF-β1 and stimulate expansion of natural T reg cells. Moreover, TGF-β1 has been shown to act as a costimulatory factor for expression of Foxp3, leading to the differentiation of CD4+CD25+ Treg cells from peripheral CD4+CD25- progeny. TGF-β-induced regulatory T cells have been termed Ti-Treg.