Please login to your online account to display your discounted pricing

TGF beta Mouse anti-Human, Biotin, Clone: eBio16TFB, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $404.55

Specifications

Antigen TGF beta
Clone eBio16TFB
Host Species Mouse
Gene Alias TGF beta
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122621

 
life technologies
LS13992381
50μg Each for $143.55

501122622

 
life technologies
LS13992385
500μg Each for $404.55
Description & Specifications

Specifications

Antigen TGF beta
Clone eBio16TFB
Host Species Mouse
Gene Alias TGF beta
Species Reactivity Human
Applications ELISA Detection
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The eBio16TFB antibody reacts with Transforming Growth Factor-beta (TGF-β) a pleiotropic cytokine, exists in five isoforms, known as TGF-ß1-5. Homologies between isoforms range from 70-80% but no homology exists to TGF-a. TGF-ß1 is ubiquitous and the most abundant form found in lymphoid organs, while other isoforms are expressed in a more restricted distribution. The biologically active state of all isoforms are disulfide-linked homodimers. The heat- and acid- stable monomeric subunits have a length of 112 amino acids. The heat- and acid- stable monomeric subunits have a length of 112 amino acids. The isoforms of TGF-β arise by proteolytic cleavage of longer precursors; the isoforms are derived from the carboxyterminal ends of these precursors. Isoforms isolated from different species are evolutionarily closely conserved and have sequence identities on the order of 98%. Mature human, porcine, simian, chicken and bovine TGF-β1 are identical and differ from mouse TGF-β1 in a single amino acid. TGF-β1 is produced in very high levels by platelets. Other cellular sources of TGF-β1 include macrophages, lymphocytes, endothelial cells, chondrocytes, and leukemic cells. TGF-β1 secretion can be induced by steroids, retinoids, EGF, NGF, vitamin D3, and IL-1. Activities of TGF-β1 include inhibition of cell growth for inhibitor for normal and transformed epithelial cells, endothelial cells, fibroblasts, neurons, and lymphoid cells and other hematopoietic cell types. TGF-β1 inhibits the proliferation of T cells and NK cells and downregulates the activities of activated macrophages. TGF-β1 blocks the anti-tumor activity of IL-2 – bearing lymphokine-activated killer (LAK) cells.

Recently, TGF-β1 has been found to have a critical role in the development of regulatory T cells. Dendritic cells exposed to tumors have been reported to secrete TGF-β1 and stimulate expansion of naturally-occurring T reg cells. Moreover, TGF-β1 has been shown to act as a costimulatory factor for expression of Foxp3, leading to the differentiation of CD4+CD25+ Treg cells from peripheral CD4+CD25- progeny. TGF-β-induced regulatory T cells have been termed Ti-Treg.