Please login to your online account to display your discounted pricing

TG-SDS Buffer, 10X Powder Pack, ULTROL™ Grade, Calbiochem™, EMD Millipore

Each pack provides an adequate amount of material to make 1L of 10X concentrate that can be diluted to a 1X solution containing 25mM Tris, 192mM glycine

$86.01 - $86.01

Chemical Identifiers

Analyte or Component Names Tris, Glycine, SDS
Formula Weight 121.14
CAS 77-86-1
Chemical Name or Material TG-SDS Buffer
Concentration or Composition (by Analyte or Components) 1X solution containing 25mM Tris, 192mM glycine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5852072PACK

 
emd millipore corporation
5852072PACK
Each for $86.01
Description & Specifications

Specifications

Formula Weight 121.14
Quantity 2 Pk.
Chemical Name or Material TG-SDS Buffer
Analyte or Component Names Tris, Glycine, SDS
CAS 77-86-1
Synonym 10X Tris-Glycine-SDS Buffer
Concentration or Composition (by Analyte or Components) 1X solution containing 25mM Tris, 192mM glycine
Solubility Information Easily soluble in cold water
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Safety and Handling

15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).