Please login to your online account to display your discounted pricing

TG 100713, Tocris™

PI 3-kinase inhibitor

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material TG 100713
Assay Percent Range >98%
Molecular Formula C12H10N6O
Formula Weight 254.25
Synonym 3-(2, 4-Diamino-6-pteridinyl)-phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

426410

 
r & d systems
426410
10mg Each for $284.81

426450

 
r & d systems
426450
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 254.25
Quantity 10mg
Chemical Name or Material TG 100713
Assay Percent Range >98%
Molecular Formula C12H10N6O
CAS 925705-73-3
Synonym 3-(2, 4-Diamino-6-pteridinyl)-phenol
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature