Please login to your online account to display your discounted pricing

TFM-4AS-1, Tocris™

Selective androgen receptor modulator (SARM)

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material TFM-4AS-1
Assay Percent Range >98%
Molecular Formula C27H33F3N2O2
Formula Weight 474.56
Synonym (1S,3aS,3bS,5aR,9aR,9bS,11aS)-6,9a,11a-Trimethyl-7-oxo-N-[2-(trifluoromethyl)phenyl]-2,3,3a,3b,4,5,5a,9b,10,11-decahydro-1H-indeno[5,4-f]quinoline-1-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

381310

 
r & d systems
RDS381310
10mg Each for $252.39

381350

 
r & d systems
RDS381350
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 474.56
Quantity 10mg
Assay Percent Range >98%
CAS 188589-61-9
Recommended Storage Store at Room Temperature
Molecular Formula C27H33F3N2O2
Chemical Name or Material TFM-4AS-1
Synonym (1S,3aS,3bS,5aR,9aR,9bS,11aS)-6,9a,11a-Trimethyl-7-oxo-N-[2-(trifluoromethyl)phenyl]-2,3,3a,3b,4,5,5a,9b,10,11-decahydro-1H-indeno[5,4-f]quinoline-1-carboxamide
Safety and Handling

Store at Room Temperature