Please login to your online account to display your discounted pricing

TFAP2B Recombinant Protein, Abnova

$622.91 - $622.91

Specifications

Cross Reactivity Human
Format Solution
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
Molecular Weight 75.24
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

89-010-897

 
abnova corporation
H00007021P0125
25μg Each for $622.91
Description & Specifications

Specifications

Cross Reactivity Human
Format Solution
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
Molecular Weight 75.24
Name Human TFAP2B Full-length ORF Recombinant Protein with GST-tag at N-terminal
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE stained with Coomassie Blue
Recombinant Yes
Source Wheat Germ (in vitro)
Storage Requirements -80°C

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array