missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Texwipe™ VersaWipe™ Wipers

For ISO Class 6 to 7 (Class 1000 to 10,000) applications

Manufacturer:  Texwipe™ TX629

 View more versions of this product

Catalog No. 19-120-2433




Description

Description

Highly absorbent, durable wipers made of a nonwoven blend of 55% cellulose and 45% polyester.

  • Blend of 55% cellulose and 45% polyester offers low extractable levels and particle count
  • Economical wiper is ideal general wiping and spill control in controlled environments
  • Compatible with solvents
  • Contain no chemical binders
  • Excellent wet and dry strength
  • Autoclavable

Order Info

9 × 9 in. wipers available in packs of 300 and case of 10/pack.

Specifications

Specifications

9 in.
280
60
For ISO Class 6 to 7 (Class 1000 to 10000) cleanroom and general wiping and spill control applications
23 cm
VersaWipe™ dry wipes
9 in.
23 cm
Yes
9 x 9 in. (23 x 23 cm)
9 in.
Cellulose, Polyester
9 in.
SDS
Documents
Product Certifications