missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Texwipe™ VersaWipe™ Wipers

For ISO Class 6 to 7 (Class 1000 to 10,000) applications

Manufacturer:  Texwipe™ TX624

 View more versions of this product

Catalog No. 19-150-0105



Description

Description

Highly absorbent, durable wipers made of a nonwoven blend of 55% cellulose and 45% polyester.

  • Blend of 55% cellulose and 45% polyester offers low extractable levels and particle count
  • Economical wiper is ideal general wiping and spill control in controlled environments
  • Compatible with solvents
  • Contain no chemical binders
  • Excellent wet and dry strength
  • Autoclavable

Order Info

9 × 9 in. wipers available in packs of 300 and case of 10/pack. 12 × 12 in. wipers available 10 bags/case (150 wipers/bag).

Specifications

Specifications

280
60
4 in.
4 in.
Cleanroom
10 cm
VersaWipe™ wipes
Yes
4 x 4 in. (10 x 10 cm)
4 in.
10 cm
4 in.
Cellulose, Polyester
Documents