missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Texwipe™ TexVantage™ Cellulose/Polyester Wipers

General-purpose, nonwoven wiper for cleaning and spill control in critical environments

$259.97 - $259.97

Specifications

Adsorbent Type 280
Autoclavable Yes
Construction Nonwoven, hydroentangled with bidirectional strength
Product Type TexVantage™ wipes
Material Cellulose, Polyester
View More Specs

Products 1
Catalog Number Mfr. No. Length (Metric) Sterility Width (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Sterility Width (Metric) Price Quantity  
18-366-218 View Documents Texwipe™
TX8932S
30.5 cm Sterile 30.5 cm Case of 500 Each for $259.97
 
Description

Description

Combine the high sorption of natural cellulose and the cleanliness and strength of polyester.

  • Nonwoven, hydroentangled construction with bidirectional strength
  • Available in standard or sterile packaging, expiration date marked on each package
  • Chemically resistant to withstanding exposure to common cleaning and disinfecting
  • Solvent-safe cleanroom packaging
  • Statistical quality control
  • Autoclavable

Sterile Features

  • Gamma irradiation to 10-6 Sterility Assurance Level, according to AAMI guidelines
  • Lot-specific information on each shipment simplifies recordkeeping

Applications:

General-purpose wiping, applications with high absorbency and low extractables requirements, aqueous spills, cleaning precision components and lab apparatus.

Specifications

Specifications

280
Nonwoven, hydroentangled with bidirectional strength
Cellulose, Polyester
Yes
TexVantage™ wipes