missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Texwipe™ Polysat™ Presaturated Polypropylene Wipers
For ISO Class 6 to 7 applications
Manufacturer: Texwipe™ TX3213
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
- Constructed from a soft, 100% meltblown polypropylene substrate
- Presaturated with 70% isopropyl alcohol/30% deionized water solution
- Low particle generation and low levels of solvent extractables
Order Info
Available conveniently packaged in a resealable FlexPack™ or bulk packed in a easy-to-carry, reclosable bag.
Specifications
9 in. | |
99 | |
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999 | |
Designed for cleaning equipment and other environmental surfaces during and following the production flow, suitable for wiping the exterior of production equipment in controlled environments | |
27.9 cm | |
PolySat™ prewetted wipes | |
1000/Cs. | |
11 in. |
22.8 cm | |
35 | |
11 x 9 in. (27.9 x 22.8 cm) | |
11 in. | |
Polypropylene | |
9 in. | |
70% Isopropyl Alcohol, 30% Deionised Water | |
Sterile |