Please login to your online account to display your discounted pricing
Texwipe™ KOGO™ TVR Cleanroom Adhesive Tapes
Ideal for clean build or cleanroom construction applications where quick tack and abrasion resistance are important
$36.63 - $53.47
Specifications
Certifications/Compliance ISO 3
Diameter (English) Outside 3 in.
Diameter (Metric) Outside 7.6cm
Adhesive Type Synthetic Rubber
Finish Matte
View More Specs
Products
Catalog Number Mfr. No. Width (English) Width (Metric) Color Quantity Price Quantity    
18-366-411
texwipe™
TVR0596YE
0.5 in. 1.3cm Yellow 96 Roll(s) Pack of 4 Each for $51.03
Case of 24 Pack for $1,157.52
18-366-412
texwipe™
TVR0596BL
0.5 in. 1.3cm Blue 96 Roll(s) Pack of 4 Each for $51.03
Case of 24 Pack for $1,157.52
18-366-414
texwipe™
TVR0596GR
0.5 in. 1.3cm Green 96 Roll(s) Pack of 4 Each for $51.03
Case of 24 Pack for $1,157.52
18-366-413
texwipe™
TVR0596WH
0.5 in. 1.3cm White 96 Roll(s) Pack of 4 Each for $51.03
Case of 24 Pack for $1,157.52
18-366-415
texwipe™
TVR0596RD
0.5 in. 1.3cm Red 96 Roll(s) Pack of 4 Each for $51.03
Case of 24 Pack for $1,157.52
18-366-446
texwipe™
TVR1048YE
1 in. 2.5cm Yellow 48 Roll(s) Pack of 2 Each for $38.12
Case of 24 Pack for $864.72
18-366-448
texwipe™
TVR1048WH
1 in. 2.5cm White 48 Roll(s) Pack of 2 Each for $38.73
Case of 24 Pack for $878.64
18-366-447
texwipe™
TVR1048BL
1 in. 2.5cm Blue 48 Roll(s) Pack of 2 Each for $38.12
Case of 24 Pack for $864.72
18-366-449
texwipe™
TVR1048GR
1 in. 2.5cm Green 48 Roll(s) Pack of 2 Each for $38.12
Case of 24 Pack for $864.72
18-366-452
texwipe™
TVR2024BL
2 in. 5.0cm Blue 24 Roll(s) Pack for $36.63
Case of 24 Pack for $830.88
18-366-450
texwipe™
TVR1048RD
1 in. 2.5cm Red 48 Roll(s) Pack of 2 Each for $38.12
Case of 24 Pack for $864.72
18-366-451
texwipe™
TVR2024YE
2 in. 5.0cm Yellow 24 Roll(s) Pack for $36.63
Case of 24 Pack for $830.88
18-366-453
texwipe™
TVR2024WH
2 in. 5.0cm White 24 Roll(s) Pack for $36.63
Case of 24 Pack for $830.88
18-366-454
texwipe™
TVR2024GR
2 in. 5.0cm Green 24 Roll(s) Pack for $36.63
Case of 24 Pack for $830.88
18-366-455
texwipe™
TVR2024RD
2 in. 5.0cm Red 24 Roll(s) Pack for $36.63
Case of 24 Pack for $830.88
18-366-458
texwipe™
TVR3016WH
3 in. 7.6cm White 16 Roll(s) Pack for $53.47
Case of 16 Pack for $831.36
18-366-457
texwipe™
TVR3016BL
3 in. 7.6cm Blue 16 Roll(s) Pack for $53.47
Case of 16 Pack for $831.36
18-366-459
texwipe™
TVR3016GR
3 in. 7.6cm Green 16 Roll(s) Pack for $53.47
Case of 16 Pack for $831.36
18-366-456
texwipe™
TVR3016YE
3 in. 7.6cm Yellow 16 Roll(s) Pack for $53.47
Case of 16 Pack for $831.36
18-366-460
texwipe™
TVR3016RD
3 in. 7.6cm Red 16 Roll(s) Pack for $53.47
Case of 16 Pack for $831.36
Description & Specifications
Specifications
Certifications/Compliance ISO 3
Diameter (English) Outside 3 in.
Diameter (Metric) Outside 7.6cm
Adhesive Type Synthetic Rubber
Finish Matte
For Use With (Application) Ideal for aisle marking, color coding, HEPA filter protection and cleanroom build
Latex Free Yes
Length (English) 36 yd.
Length (Metric) 33m
Material PVC
Temperature Range (English) -20 to 176°F
Temperature Range (Metric) -29 to 80°C
Thickness (English) 6mil
  • Bonds quickly to irregular surfaces such as duct work or visqueen sheets used for temporary walls
  • Matte finish is easy to write on with a pen for labeling
  • Good for color coding or aisle marking in cleanrooms
  • PVC backing material with synthetic rubber adhesive
  • Roll length: 33m (36 yd.)
  • Various colors and widths available
Packaged in press-and-seal cleanroom bags
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.