Please login to your online account to display your discounted pricing
Texwipe™ Cleanroom Tape
Good chemical and thermal resistance
$721.90 - $895.75
Products
Catalog Number Mfr. No. Price Quantity    
296356761
texwipe™
AT2024WH
Case of 24 Each for $895.75
296356757
advanced laminated
AT1048WH
Case of 48 Each for $727.13
296356760
advanced laminated
AT1048CL
Case of 48 Each for $721.90
Description & Specifications
  • Low-particulate, contaminant-free tape features a unique adhesive that offers superior bonding properties
  • Removes cleanly from most surfaces, leaving little or no residue
  • Provides a superior moisture barrier
  • Heat sealed in an ISO Class 5 cleanroom
  • Cleaned and wound on a 3 in. dia. (7.6cm) particulate-free polyethylene core
Rolls are 36 yd. long (33m) and come in widths from 0.5 to 2 in. (1.3 to 5.1cm). Double bagged. Contact your Fisher Safety Customer Service Representative for availability and ordering information on additional colors and sizes.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.