Texwipe™ Cleanroom Spiral Notebooks
For note-taking and data recording
Manufacturer:  Texwipe™ TX5742
 View more versions of this product
Catalog No. 19-130-5044
Description
Description
  • High-density polyethylene covers, plastic spirals, and durable TexWrite™ latex-impregnated cleanroom paper
  • White, college-ruled pages printed with IPA-resistant, low-sodium ink for reduced ionic contamination
  • Notebooks lie flat; pages rotate freely for ease of use
  • 100 writable pages (50 sheets)
Specifications
Specifications
Paper
3 x 5 in. (76 x 127mm)
3 in.
22 lb.
5 in.
HDPE
White
For recording notebook, record keeping, data transfer and notes
76mm
9.97kg
127mm
Cleanroom Spiral Notebook
Documents