missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Texas Instruments™ TI-Nspire™ CX CAS Handheld — Teacher's Bundle and Pack

A virtual and real-world way to explore science.

$376.65 - $376.65

Specifications

Battery Type Nspire rechargeable
Grade Middle to High School
Keypad Touchpad navigation
Memory 64MB
Resolution 125dpi
View More Specs

Includes:
  • Teacher Bundle: One handheld, single-license TI-Nspire and CAS Teacher Software, rechargeable battery, wall adapter, USB unit-to-unit cable, USB unit-to-computer cable
  • Teacher Pack: Ten handhelds with rechargeable batteries installed, 90-day trial of TI-Nspire and CAS Teacher Software, ten USB cables, five unit-to-unit cables, one manual, docking station and poster


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
S02282ND Fisher Science Education
N3CAS/CBX/2L1/B
One handheld, single license TI-Nspire and CAS Teacher Software, rechargeable battery, wall adapter, USB unit to unit cable, USB unit to computer cable Teacher Bundle Each for $376.65
Specifications

Description

The perfect solution for the classroom, lab and field. Explore dynamic simulations and collect data with Vernier™ sensors.

  • Explore hundreds of free TI-Nspire biology, chemistry, physics and earth science lessons and simulations from middle to high school grades at www.sciencenspired.com
  • Instantly turn the TI-Nspire CX handheld into a data collection device by attaching a Vernier EasyLink™ USB interface and sensor
  • For advanced data collection, use the TI-Nspire Lab Cradle (sold separately); supports simultaneous use of up to five Vernier sensors
  • Use digital images or your own photos to connect science concepts with real-world scenarios
  • Touchpad navigation works more like a laptop computer
  • CAS (Computer Algebra System) allows for symbolic and numeric calculations such as scientific unit cancellation or combination
  • Approved for standardized tests — SAT™, PSAT/NMSQT™ and AP exams
  • Screen size: 320 × 240 pixels (3.2 in. [8.1cm] diagonal)
  • Screen resolution: 125 dpi; 16-bit color
  • Memory: 100MB storage memory/64MB operating memory

Order Info

Powered by the TI-Nspire rechargeable battery (included) that lasts up to two weeks of normal use on a single charge. No alkaline batteries needed.

Description & Specifications

Specifications

Nspire rechargeable
Touchpad navigation
125dpi
Middle to High School
64MB
320 x 240