Please login to your online account to display your discounted pricing

Tetrindole mesylate, Tocris

MAO-A inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Tetrindole mesylate
Assay Percent Range >99%
Molecular Formula C20H26N2·CH3SO3H
Formula Weight 390.54
Synonym 2,3,3a,4,5,6-Hexahydro-8-cyclohexyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

072350

 
r & d systems
072350
50mg Each for $682.67

072310

 
r & d systems
072310
10mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 390.54
Assay Percent Range >99%
Chemical Name or Material Tetrindole mesylate
CAS 170964-68-8
Recommended Storage Store at Room Temperature
Molecular Formula C20H26N2·CH3SO3H
Synonym 2,3,3a,4,5,6-Hexahydro-8-cyclohexyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
Safety and Handling

Store at Room Temperature