Please login to your online account to display your discounted pricing

Tetrindole mesylate, Tocris


MAO-A inhibitor

Quantity 50mg

Manufacturer: r & d systems  072350

 View more versions of this product

Catalog No. 072350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 390.54
Chemical Name or Material Tetrindole mesylate
Assay Percent Range >99%
Molecular Formula C20H26N2·CH3SO3H
CAS 170964-68-8
Synonym 2,3,3a,4,5,6-Hexahydro-8-cyclohexyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Tetrindole mesylate
Assay Percent Range >99%
Molecular Formula C20H26N2·CH3SO3H
Formula Weight 390.54
Synonym 2,3,3a,4,5,6-Hexahydro-8-cyclohexyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
Accessories

Store at Room Temperature