missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™, ACROS Organics™

Manufacturer:  Acros Organics AC432991000

 View more versions of this product

Catalog No. AC432991000



Specifications

Specifications

Tetrazole
AcroSeal™ Glass Bottle
Cream to Orange
288-94-8,75-05-8
1h-tetrazole, tetrazole, tetraazacyclopentadiene, 1-h-tetrazole, 1h-1,2,3,4-tetrazole, 1 h-tetrazole, 1h-tetrazole solution, 2h-1,2,3,4-tetrazole, racemic tetrazole, 1h-tetraazole
C1=NNN=N1
70.055
CHEBI:33193
Crystalline Powder
0.8000g/mL
0.3% max.
100mL
CH2N4
KJUGUADJHNHALS-UHFFFAOYSA-N
2H-tetrazole
67519
70.05

Chemical Identifiers

288-94-8
70.055
1h-tetrazole, tetrazole, tetraazacyclopentadiene, 1-h-tetrazole, 1h-1,2,3,4-tetrazole, 1 h-tetrazole, 1h-tetrazole solution, 2h-1,2,3,4-tetrazole, racemic tetrazole, 1h-tetraazole
CHEBI:33193
C1=NNN=N1
CH2N4
KJUGUADJHNHALS-UHFFFAOYSA-N
67519
2H-tetrazole
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapour.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents