missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetravinyl tin, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC321080250

 View more versions of this product

Catalog No. AC321080250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Tetravinyl tin
40°C
Glass bottle
1.24
Colorless to Yellow
1112-56-7
C8H12Sn
MFCD00008608
MZIYQMVHASXABC-UHFFFAOYSA-N
tetrakis(ethenyl)stannane
66189
Liquid
1.2400g/mL
Authentic
1.5000 to 1.503
160°C to 163°C
25mL
94% min. (GC)
Sn(CH=CH2)4
sn ch=ch2 4, stannane, tetraethenyl, stannane, tetravinyl, stannane,tetravinyl, tetraethenylstannane, tetravinyl tin, tetravinylstannane, tetravinyltin, tetravinyltin iv, tin tetravinyl
C=C[Sn](C=C)(C=C)C=C
226.88
226.88
95%

Chemical Identifiers

1112-56-7
226.88
MZIYQMVHASXABC-UHFFFAOYSA-N
66189
C=C[Sn](C=C)(C=C)C=C
C8H12Sn
MFCD00008608
sn ch=ch2 4, stannane, tetraethenyl, stannane, tetravinyl, stannane,tetravinyl, tetraethenylstannane, tetravinyl tin, tetravinylstannane, tetravinyltin, tetravinyltin iv, tin tetravinyl
tetrakis(ethenyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-193-6

SDS Documents