missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetratetracontane 97.0+%, TCI America™
$63.38 - $275.87
Chemical Identifiers
| CAS | 7098-22-8 |
|---|---|
| Molecular Formula | C44H90 |
| Molecular Weight (g/mol) | 619.204 |
| MDL Number | MFCD00015268 |
| InChI Key | KMXFZRSJMDYPPG-UHFFFAOYSA-N |
| Synonym | n-tetratetracontane, tetratetracontane, analytical standard, acmc-209ogu, tetratetracontane, tetratetracontane, purum gc |
| PubChem CID | 23494 |
| ChEBI | CHEBI:84289 |
| IUPAC Name | tetratetracontane |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T0629100MG
|
TCI America
T0629100MG |
100 mg |
Each for $63.38
|
|
|||||
|
T06291G
|
TCI America
T06291G |
1 g |
Each for $275.87
|
|
|||||
Chemical Identifiers
| 7098-22-8 | |
| 619.204 | |
| KMXFZRSJMDYPPG-UHFFFAOYSA-N | |
| 23494 | |
| tetratetracontane |
| C44H90 | |
| MFCD00015268 | |
| n-tetratetracontane, tetratetracontane, analytical standard, acmc-209ogu, tetratetracontane, tetratetracontane, purum gc | |
| CHEBI:84289 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
Specifications
| 7098-22-8 | |
| White | |
| MFCD00015268 | |
| n-tetratetracontane, tetratetracontane, analytical standard, acmc-209ogu, tetratetracontane, tetratetracontane, purum gc | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC | |
| 619.204 | |
| CHEBI:84289 | |
| ≥97.0% (GC) | |
| Tetratetracontane |
| 86°C | |
| C44H90 | |
| 100 mg | |
| KMXFZRSJMDYPPG-UHFFFAOYSA-N | |
| tetratetracontane | |
| 23494 | |
| 619.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes